Orders LinkContactUsLinkComponent
Paterson® Auto Load Reel , Electron Microscopy Sciences
Paterson® Auto Load Reel , Electron Microscopy Sciences
Paterson Auto Load Reel
Catalog # 100502-862
Supplier #:  74338-01
Paterson® Auto Load Reel , Electron Microscopy Sciences
Catalog # 100502-862
Paterson® Auto Load Reel , Electron Microscopy Sciences
Paterson® Auto Load Reel , Electron Microscopy Sciences
Paterson Auto Load Reel
Price
$25.53
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed for use in the Paterson Super System 4 tank, the Auto Load Reel allows thru free flow of solutions. Paterson Reels have a smooth finish and use the unique twin ball ratchet system. Film loading just couldn’t be easier.
Specifications
  • Description:
    Paterson Auto Load Reel