View-Pack™ Microscope Slide Holder With 3-Ring Binder, Electron Microscopy Sciences
Product Details & Documents
For the orderly visual display and safe transport of microscope slides.
This loose-leaf binder has ten vinyl View-Pack pages (hold 160 slides). Each page holds 16 slides 75 x 25 mm (3 x 1") in individual pockets with a white back and clear front. A flap in the center keeps slides from falling out. Simply fold the page at the center crease to retrieve slides. The 22 x 27 cm (8 1/2 x 10 1/2") View-Pack pages comes with a standard 23 x 30 cm (9 x 12") 3-ring binder.
Specifications
- Description:View Pak 3-Ring Binder