Tray Set with Plastic Boxes, Electron Microscopy Sciences
Product Details & Documents
A compact, solid fiberboard tray box makes instituting a plastic box/sample storage system more convenient and efficient.
- Available with or without accompanying plastic boxes
To form a tray set, use plastic boxes for specimen storage, and then organize boxes within the fiberboard tray Box. These sets eliminates misplacement or inadvertent sample loss and allows for rapid retrieval of specimens.
Specifications
- Description:Tray Set With Plastic Boxes
- Size:25 Box Set