Diamond Points, Electron Microscopy Sciences
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
- Ø:7.3 mm
- Description:Diamond Points, Tapered
- Shape:Wheel