Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Formaldehyde/Zinc Fixative, Electron Microscopy Science

Formaldehyde/Zinc Fixative, Electron Microscopy Science

Formaldehyde/Zinc Fixative, 1 L
Catalog # 100496-486
Minority or Woman-Owned Business Enterprise
Supplier #:  15675-01
Formaldehyde/Zinc Fixative, Electron Microscopy Science
Catalog # 100496-486
Minority or Woman-Owned Business Enterprise
CAS Number:  
Formaldehyde/Zinc Fixative, Electron Microscopy Science

Formaldehyde/Zinc Fixative, 1 L

CAS Number:  
Price
$46.38
Log in for availability
Each (1l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Formaldehyde/zinc fixative is ready-to-use solution. Can be used with automated tissue processors.

  • Preserves fine nuclear details
  • Good for immunostaining
  • Contains no mercury

Works well with standard and special stains because zinc is a stain enhancing mordant.

Specifications

Specifications

  • Description:
    Formaldehyde/Zinc Fixative
  • Size:
    1 L
  • Cat. No.:
    100496-486
  • Storage Temperature:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.