Orders LinkContactUsLinkComponent
Micrometer, Electron Microscopy Sciences
Micrometer, Electron Microscopy Sciences
Micrometer, Model A
Catalog # 100496-128
Supplier #:  62060
Micrometer, Electron Microscopy Sciences
Catalog # 100496-128
Micrometer, Electron Microscopy Sciences
Micrometer, Electron Microscopy Sciences
Micrometer, Model A
Price
$83.40
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Metric and English scale micrometers with friction stop; comes in a padded plastic case.
Model A: Calibrated to read 0 to 25 mm, in 0.01 mm divisions.
Model B: Calibrated to read 0 to 1", in 0.001" divisions.
Specifications
  • Description:
    Micrometer, Model A