Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Thallium Grain
Catalog # 100495-226
Supplier #:  80075-TL
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Catalog # 100495-226
CAS Number:  
MAC X-ray Microanalysis Individual Standards, Grains, Electron Microscopy Sciences
Thallium Grain
CAS Number:  
Price
$534.39
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with a location map for standard identification. Grains of standard materials are available and they measure approximately 2 x 2 mm in size. They are supplied with fully authenticated certificates of analysis.
Specifications
Specifications
  • Description:
    Thallium Grain