Orders LinkContactUsLinkComponent
Negative Carriers, Electron Microscopy Sciences
Insert for 35 mm
Catalog # 100494-088
Negative Carriers, Electron Microscopy Sciences
Catalog # 100494-088
Supplier:  
Negative Carriers, Electron Microscopy Sciences
Negative Carriers, Electron Microscopy Sciences
Insert for 35 mm
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts
Glassless, sandwich type negative carriers are rofessional quality, with black, anodized aluminum construction. Hinged for quick handling.
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts. Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Specifications
  • Description:
    Insert for 35 mm