Orders LinkContactUsLinkComponent
Scotch Conductive Tapes, Electron Microscopy Sciences
Conductive Aluminum Tape, ¼"
  100492-336
Scotch Conductive Tapes, Electron Microscopy Sciences
  100492-336
Scotch Conductive Tapes, Electron Microscopy Sciences
Scotch Conductive Tapes, Electron Microscopy Sciences
Conductive Aluminum Tape, ¼"
$56.68
Each
Copper and aluminum conductive tapes with continuous long term usage up to 155 °C
  • Functional days-weeks usage up to 175 °C
  • Copper double sided available
We offer two types of foil: Aluminum foil tape (Scotch 1170) and copper foil tape (Scotch 1181) with conductive adhesive. 3” core, 1/4" (6.35 mm) width×54 ft (16.5 m) long.
  • Width:
    ¼"
  • Description:
    Conductive Aluminum Tape