Orders LinkContactUsLinkComponent
Darkroom Thermometer, Electron Microscopy Sciences
Luminescent Thermometer, Model B
Catalog # 100492-194
 :  74389
Darkroom Thermometer, Electron Microscopy Sciences
Catalog # 100492-194
Darkroom Thermometer, Electron Microscopy Sciences
Darkroom Thermometer, Electron Microscopy Sciences
Luminescent Thermometer, Model B
Price
$72.53
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Model A comes with 1 3/4" dial with a 6" stem.
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2°.
Model A comes with 1 3/4" dial with a 6" stem.
Model B comes with 2 1/2" luminescent dial with a 8" stem, 1/2° graduations and a calibration adjustment wrench.
Specifications
  • Description:
    Luminescent Thermometer, Model B