Orders LinkContactUsLinkComponent
Hinged Lid Containers, PE, Electron Microscopy Sciences
Hinged Lid Containers, PE, Electron Microscopy Sciences
0.25 oz., Hinged Lid Containers, ½", 11/2"
Catalog # 100490-462
Supplier #:  64238-01
Hinged Lid Containers, PE, Electron Microscopy Sciences
Catalog # 100490-462
Hinged Lid Containers, PE, Electron Microscopy Sciences
Hinged Lid Containers, PE, Electron Microscopy Sciences
0.25 oz., Hinged Lid Containers, ½", 11/2"
Price
$117.64
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These hinged lid containers are ideal for use just about anywhere.
  • Secure snap closure with hinged lid that eliminates lost caps
  • Offer air and moisture resistance to contents
  • Manufactured from flexible FD approvable polyethylene resin
Specifications
  • Ø:
    11/2"
  • Capacity:
    0.25 oz.
  • Description:
    Hinged Lid Containers
  • Height:
    ½"