Ampoule Cracker, Electron Microscopy Sciences
Product Details & Documents
The Ampoule Cracker™ is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules.
Simply slip the Ampoule Cracker™ over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Specifications
- Description:Ampoule Breaker
- For:1 - 4 ml ampoules