Orders LinkContactUsLinkComponent
Syringes Disposable, Electron Microscopy Sciences
20 ml, Luerslip Eccentric
  100488-484
Syringes Disposable, Electron Microscopy Sciences
  100488-484
Syringes Disposable, Electron Microscopy Sciences
Syringes Disposable, Electron Microscopy Sciences
20 ml, Luerslip Eccentric
$130.59
Pack of 100
These high quality disposable rubber piston syringes offer substantial savings.
  • Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
  • Syringes are ETO gas sterilized, latex and pyrogen free
  • Bold graduations are easily read and resist wipe-off
 : All syringes are packaged in individual sterile strip packs.
  • Capacity:
    20 ml
  • Tip style:
    Luerslip Eccentric