Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
Biological Set with 32 mm dia Block
Catalog # 100488-092
Supplier #:  80072-32
MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
Catalog # 100488-092
CAS Number:  
MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
MAC X-ray Microanalysis Standards Biological Set, Electron Microscopy Sciences
Biological Set with 32 mm dia Block
CAS Number:  
Price
$5,165.33
Log in for availability
Kit of 18
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems
A set of 18 standards, a Faraday Cup included. Mounted in 25 or 32 mm diameter brass block. Includes Al, BaF2, BaSO4, Bi, BN, C , CaCO3, CaSO4, FeS2, InP, KBr, KCl, MgO, NaCl, Se, SiO2, Ti, V.
All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.
Specifications
Specifications
  • Description:
    Biological Set with 32 mm dia Block