Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Universal® Wall Files

Universal® Wall Files

Expandable single pocket with add-on pocket, Three-pocket starter sets (UNV53682), Clear, Letter, 1, 12¾", 1¾", 7"
Catalog # 500010-821
Supplier:   Janitorial Supplies
Supplier #:  UNV53692
Universal® Wall Files
Catalog # 500010-821
Universal® Wall Files
Universal® Wall Files

Expandable single pocket with add-on pocket, Three-pocket starter sets (UNV53682), Clear, Letter, 1, 12¾", 1¾", 7"

Price
$9.41
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Expandable system lets you customize your wall filing system

Simply add pockets as your organization needs grow. Mounting hardware included. Sturdy plastic is tough enough for active environments.

Specifications
  • For Use With:
    Three-pocket starter sets (UNV53682)
  • Color:
    Clear
  • Height:
    7"
  • Size:
    Letter
  • Depth:
    1¾"
  • Type:
    Expandable single pocket with add-on pocket
  • Width:
    12¾"
  • No. of pockets:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.