Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
iScript™ Select cDNA Synthesis Kit, Bio-Rad

iScript™ Select cDNA Synthesis Kit, Bio-Rad

iScript™ Select cDNA Synthesis Kit, 100× 20 µl
Catalog # 101645-414
Marketsource
Supplier:   Bio-Rad
Supplier #:  1708897
iScript™ Select cDNA Synthesis Kit, Bio-Rad
Catalog # 101645-414
Supplier:  Bio-Rad
Marketsource
CAS Number:  
iScript™ Select cDNA Synthesis Kit, Bio-Rad
iScript™ Select cDNA Synthesis Kit, Bio-Rad

iScript™ Select cDNA Synthesis Kit, 100× 20 µl

CAS Number:  
Price
$1,061.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The iScript™ cDNA Synthesis Kit is a sensitive and easy-to-use first-strand cDNA synthesis kit for gene expression analysis using RT-qPCR or conventional RT-PCR

  • Dilute RNA samples
  • Format allows up to 15 µl of input RNA in a 20 µL reaction
  • Low-level genes and RNA conservation
  • Broad linear dynamic range of total input RNA (1 µg–1 pg)
  • RNase A inhibitor for RNA protection during setup and reaction
  • Blend of oligo(dT) and random primers
  • Prevent 5' and 3' bias, and allow primers designed for any region

This two-tube kit is optimized for reproducible, unbiased representation over a broad dynamic range, with a fast 40 min protocol.

Ordering information: Each kit contains 5x reaction mix, iScript™ Reverse Transcriptase (RNase H+ MMLV), and nuclease-free water. Guaranteed for 12 months at —20°C in a constant temperature freezer (multiple freeze thaws not recommended).

Documents

Specifications

Specifications

  • Description:
    iScript™ Select cDNA Synthesis Kit
  • No. of reactions:
    100× 20 µl

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.