Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Fixatives, Prepared, Electron Microscopy Sciences

Fixatives, Prepared, Electron Microscopy Sciences

2.5% Glutaraldehyde in 0.1 M Sodium Cacodylate Buffer (pH 7.4)
Catalog # 100503-956
Supplier #:  15960
Fixatives, Prepared, Electron Microscopy Sciences
Catalog # 100503-956
CAS Number:  
Fixatives, Prepared, Electron Microscopy Sciences
Fixatives, Prepared, Electron Microscopy Sciences

2.5% Glutaraldehyde in 0.1 M Sodium Cacodylate Buffer (pH 7.4)

CAS Number:  
Price
$86.44
Log in for availability
Each (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.

Certifications: Certificates of analysis are available.

Documents

Specifications

Specifications

  • Description:
    2.5% Glutaraldehyde in 0.1 M Sodium Cacodylate Buffer (pH 7.4)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.