Square and Grid Scales, Electron Microscopy Sciences
Product Details & Documents
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
Specifications
- Description:NE34 Graticule
- Size:19 mm
- Divisions:Grid (net) 0.1 mm squares