Orders LinkContactUsLinkComponent
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
1/10 Hp Flex Shaft Machine Set, 110 V
Catalog # 100497-884
Supplier #:  62131-10
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Catalog # 100497-884
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
1/10 Hp Flex Shaft Machine Set, 110 V
Price
$356.26
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Specifications
  • Description:
    1/10 Hp Flex Shaft Machine Set
  • Voltage:
    110 V
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Standard Foot Speed Control
    Standard Foot Speed Control
    100497-914
    Log in for availability
    Each - $126.19
Product Information
  • Catalog #
    Availability
    Price
  • Standard Foot Speed Control
    Standard Foot Speed Control
    100502-480
    Log in for availability
    Each - $59.71
Product Information
  • Catalog #
    Availability
    Price
  • Probe Handle
    Probe Handle
    100497-882
    Log in for availability
    Each - $29.87
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Flex Shaft
    Replacement Flex Shaft
    100497-888
    Log in for availability
    Each - $77.59
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Sheath
    Replacement Sheath
    100497-890
    Log in for availability
    Each - $67.94
Product Information
  • Catalog #
    Availability
    Price
  • 13-Piece Assortment Set
    13-Piece Assortment Set
    100497-916
    Log in for availability
    Each - $44.53
Product Information
  • Catalog #
    Availability
    Price
  • 26-Piece Assortment Set
    26-Piece Assortment Set
    100497-918
    Log in for availability
    Each - $104.74
Product Information
  • Catalog #
    Availability
    Price
  • Flex Shaft Stand
    Flex Shaft Stand
    100502-482
    Log in for availability
    Each - $64.58