Orders LinkContactUsLinkComponent
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Sampling Tool 1.0
Catalog # 100492-812
Supplier #:  69036-10
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Catalog # 100492-812
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Sampling Tool 1.0
Price
$65.65
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Harris Uni-Core™ is a disposable multi-purpose sampling tool for soft substrates. Made from polypropylene, including a hollow body with built-in tool steel razor sharp cutting tip, with a plunger for the expelling of the punched sample.
  • Individually wrapped
  • Sterilized with ethylene oxide
  • Reuse by rinsing the tip with ethanol and autoclaving for 20 minutes at 250 °F/15 psi
Specifications
  • Description:
    Sampling Tool 1.0