Orders LinkContactUsLinkComponent
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Light Microscopy Perfect Loop Only
Catalog # 100491-688
Supplier #:  70941
Perfect Loop, Electron Microscopy Sciences
Catalog # 100491-688
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Light Microscopy Perfect Loop Only
Price
$238.47
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
Specifications
  • Description:
    Light Microscopy Perfect Loop Only