Orders LinkContactUsLinkComponent
Pocket Measuring Microscopes, Electron Microscopy Sciences
Pocket Measuring Microscope
  100491-448
Pocket Measuring Microscopes, Electron Microscopy Sciences
  100491-448
Pocket Measuring Microscopes, Electron Microscopy Sciences
Pocket Measuring Microscopes, Electron Microscopy Sciences
Pocket Measuring Microscope
$147.15
Each
As small as a fountain pen, the pocket measuring microscope has a standard scale is incorporated in its lens barrel, giving an effecient brightness and excellent definition. It can be focused in the same way as a conventional microscope.
  • 25x magnification has 3.30 mm field of view, 3 mm measuring range and minimal scale division of 0.005
  • 50x magnification has 2.00 mm field of view, 1.6 mm measuring range and minimal scale division of 0.02
Dimensions: 12.40 x 127 mm
  • Magnification:
    25×
  • Description:
    Pocket Measuring Microscope