Orders LinkContactUsLinkComponent
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial-A-Grid Box
Catalog # 100490-976
Supplier #:  71148-10
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Catalog # 100490-976
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Dial-A-Grid Box
Price
$887.51
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A two tone color coded plastic box with insert which has 24 letter-labeled crossed slots, where the grids can be stored. A rotating protection plate covers the slots and allows for exposure of one grid at a time. Measures L x W x H: 57 x 45 x 12.5 mm (2 1/3 x 1 3/4 x 1/2").
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Specifications
  • Description:
    Dial-A-Grid Box