Orders LinkContactUsLinkComponent
Histology Containers, Electron Microscopy Sciences
Histology Containers, Electron Microscopy Sciences
Opaque
Catalog # 100490-454
Supplier #:  64233-22
Histology Containers, Electron Microscopy Sciences
Catalog # 100490-454
Histology Containers, Electron Microscopy Sciences
Histology Containers, Electron Microscopy Sciences
Opaque
Price
$352.82
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing
  • Designed for storage and transport of histology specimens
  • Made from durable polypropylene (except 8L size which is made from polyethylene).
  • Leak proof seal caps.
The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.
All of the containers (except the 5 mL size) are graduated on three sides.
Specifications
  • Color:
    Opaque