Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences

Graduated Pipettes, Polypropylene, Electron Microscopy Sciences

Volumetric, 25 ml
Catalog # 100489-206
Supplier #:  70952-25
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Catalog # 100489-206
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences

Volumetric, 25 ml

Price
$243.88
Log in for availability
Pack of 3
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Made from translucent polypropylene.

  • Resistant to breakage
  • Blue graduations, ring mark and inscriptions

Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.

NOTE: Exposure to temperatures above 60 °C may cause volume changes.

Specifications
  • Capacity:
    25 ml
  • Description:
    Volumetric

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.