Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SRT® SEC HPLC Column, Sepax Technologies

SRT® SEC HPLC Column, Sepax Technologies

Analytical, SRT® SEC, 150 Å, 5 µm, 4.60 mm, 300 mm, PEEK
Catalog # 101450-342
Marketsource
Supplier:   Sepax Technologies
Supplier #:  215150P-4630
SRT® SEC HPLC Column, Sepax Technologies
Catalog # 101450-342
Supplier:  Sepax Technologies
Marketsource
SRT® SEC HPLC Column, Sepax Technologies

Analytical, SRT® SEC, 150 Å, 5 µm, 4.60 mm, 300 mm, PEEK

Price
$2,887.69
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

SRT® SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.

  • High separation efficiency and resolution
  • High capacity
  • High lot-to-lot reproducibility
  • High protein recovery with intact biological activity
  • Negligible non-specific interactions

SRT® uses stand-up monolayer on porous silica as oppose to SRT®-C of lay-down monolayer, offers ideal phase for monoclonal antibodies, proteins, peptides, nucleic acids, oligonucleotides, virus and water-soluble polymers.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4.60 mm
  • Particle Size:
    5 µm
  • Pore size:
    150 Å
  • Length:
    300 mm
  • Packing material:
    PEEK
  • Phase:
    SRT® SEC

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.