Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Needle and Knife Interchangeable Handle, Electron Microscopy Sciences

Needle and Knife Interchangeable Handle, Electron Microscopy Sciences

Handle
Catalog # 101413-604
Supplier #:  72953-HD
Needle and Knife Interchangeable Handle, Electron Microscopy Sciences
Catalog # 101413-604
Needle and Knife Interchangeable Handle, Electron Microscopy Sciences
Needle and Knife Interchangeable Handle, Electron Microscopy Sciences

Handle

Price
$12.52
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Handle L x Diameter: 92 x 6 mm (3 5/8 x 1/4")

Made from stainless steel, pin-vise type Handle, which accepts either dissecting needles or a knife.

Handle L x Diameter: 92 x 6 mm (3 5/8 x 1/4")
Needle: available with straight or bent, 60 mm (2 1/2") long
Knife: spear type with single or double edge, 75 mm (3") long

Specifications
  • Description:
    Handle

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.