Orders LinkContactUsLinkComponent
Serum Bottles, Electron Microscopy Sciences
Serum Bottles, Electron Microscopy Sciences
5 ml, Serum Bottles, Amber
Catalog # 101412-402
Supplier #:  60308-05
Serum Bottles, Electron Microscopy Sciences
Catalog # 101412-402
Serum Bottles, Electron Microscopy Sciences
Serum Bottles, Electron Microscopy Sciences
5 ml, Serum Bottles, Amber
Price
$981.27
Log in for availability
Case of 288
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Clear bottles are manufactured from '400' borosilicate molded glass. Amber bottles are manufactured from '500' borosilicate molded glass.
Certifications: Meets USP Type I standards.
Specifications
  • Capacity:
    5 ml
  • Color:
    Amber
  • Description:
    Serum Bottles