Orders LinkContactUsLinkComponent
Hall’s Method for Bilirubin, Electron Microscopy Sciences
Van Gieson Solution, 500 ml
  101411-438
 :  26611-02
Hall’s Method for Bilirubin, Electron Microscopy Sciences
  101411-438
 :  
Hall’s Method for Bilirubin, Electron Microscopy Sciences
Van Gieson Solution, 500 ml
 :  
$69.46
Each (500ml)
Hall’s Method for Bilirubin
Stain results - Biliverdin: Green, Collagen: Red, Muscle: Yellow.
  • Description:
    Van Gieson Solution
  • Size:
    500 ml