Benchtop Mat, Electron Microscopy Sciences
Product Details & Documents
A flexible polyethylene film backing which has a highly absorbent upper surface which traps water, dyes, resins, solvents, and radioactive solutions.
- Sheets can be rolled up and discarded after use
- Ideal for daily use in EM labs
Benchtop mat comes in a roll 50.8 cm (20") wide and 15.2 m (50') long in a dispenser carton, or a cut size of 20×24" sheets.
Specifications
- Description:Benchtop Mat