Orders LinkContactUsLinkComponent
Bright-Line Counting Chamber, Electron Microscopy Sciences
Phase Chamber Set without Cover Glass (1475)
  100502-518
 :  63514-20
Bright-Line Counting Chamber, Electron Microscopy Sciences
  100502-518
Bright-Line Counting Chamber, Electron Microscopy Sciences
Bright-Line Counting Chamber, Electron Microscopy Sciences
Phase Chamber Set without Cover Glass (1475)
$798.53
Each
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
 : The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.
  • Description:
    Phase Chamber Set without Cover Glass (1475)