Syringes Disposable, Electron Microscopy Sciences
Product Details & Documents
These high quality disposable rubber piston syringes offer substantial savings.
- Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
- Syringes are ETO gas sterilized, latex and pyrogen free
- Bold graduations are easily read and resist wipe-off
: All syringes are packaged in individual sterile strip packs.
Specifications
- Capacity:1 ml
- Tip style:Tuberculin
- Needle length:5/8"
- Needle gauge:25 G