Orders LinkContactUsLinkComponent
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Paper Points, Selection 1
Catalog # 100500-760
Supplier #:  71011-01
Absorbant Paper Points, Electron Microscopy Sciences
Catalog # 100500-760
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Paper Points, Selection 1
Price
$44.38
Log in for availability
Box of 200
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids. Available in two assortment sizes. The bigger the number the larger the point.
Specifications
  • Description:
    Paper Points, Selection 1