Orders LinkContactUsLinkComponent
Chalkley Point Array Graticules, Electron Microscopy Sciences
Chalkley Point Array Graticules, Electron Microscopy Sciences
NG52 Graticule, 21 mm
Catalog # 100499-288
Supplier #:  68034-21
Chalkley Point Array Graticules, Electron Microscopy Sciences
Catalog # 100499-288
Chalkley Point Array Graticules, Electron Microscopy Sciences
Chalkley Point Array Graticules, Electron Microscopy Sciences
NG52 Graticule, 21 mm
Price
$481.95
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This is used to quickly determine the relationship of components to each other using random sampling.
Curtis gives an example of its application, where a researcher may want to see whether or not a certain drug affects the volume proportion of cell types in a given organ. With this graticule the proportion of points lying over the image of one type of component is statistically proportional to the area occupied by that component. The 25 points of the array are placed over the field of view at random, so that a comparison can be made between the number of points touching the one type of component, with number touching the other type of component in each viewing. A series of observations will yield an increasingly accurate ratio of the comparative incidence of each type of particle.
Specifications
  • Description:
    NG52 Graticule
  • Size:
    21 mm