Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Product Details & Documents
The innoculating loops are made from durable, heat resistant brass with a fiber glass insulating sleeve over the gripping area. The threaded collet type collar holds all wire gauge.
Specifications
- Description:Loop and Needle Holder
- Length:152 mm (6")