Fine Diamond Scriber, Electron Microscopy Sciences
Product Details & Documents
This diamond scriber is the ultimate scribing instrument offering the most precision for the most delicate applications.
- Fine scribing under the microscope - for silicon wafers and glass coverslips
- Precision scribing and repairing - for thin film circuits and microcircuits, and more
It is available in straight or bent configurations.The integrated 6.35 mm anodized hexagonal (non-rolling) aluminum handle has a positive finger grip in the form of grooves for easy control and nonslippage. Each diamond scriber is packaged in a protective plastic tube.
Specifications
- Description:Diamond scriber, bent, 60° point angle
- Diameter:0.25 mm