Orders LinkContactUsLinkComponent
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Glennel®, Diamond Suspension, Electron Microscopy Sciences
1.0 µm
Catalog # 100497-350
Supplier #:  50372-40
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Catalog # 100497-350
Glennel®, Diamond Suspension, Electron Microscopy Sciences
Glennel®, Diamond Suspension, Electron Microscopy Sciences
1.0 µm
Price
$127.53
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Glennel® Diamond Suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring.
  • A versatile, water based suspension designed for most production applications
  • Excellent lubrication
  • Uniform fast lapping action
  • Maximum cutting action
  • Compatible with all common extenders
The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators.
Packaging: All Diamond Suspensions are packed in 400 g bottles and have Medium Concentration prepared with Premium Polycrystalline Diamond in either Propylene Glycol (Oil based) or Water based formulas.
Specifications
  • Size:
    1.0 µm