Talking Timer, Electron Microscopy Sciences
Product Details & Documents
A unique timer featuring a clear human voice that makes announcements which can be heard from any where within the lab.
- Counts up or down to 24 hours
- Has a built in stand for bench top use, clips to a pocket
- Can be stick to any metal surface with a magnetic back
Other features include a large ³/₄" display with icons to indicate various modes, such as voice off/on, count-down/count-up and more. Recalls the previous count-down setting for immediate reuse and memorizes other frequently used setting for reuse at any time.
In the count-down mode, Talking Timer will announce the remaining time every hour, In the final hour, it will announce the time remaining every 10 minutes, every minute in the final 10 minutes, and every second in the final 10 seconds. A choice of 6 distinctive final alarm sounds make it easy to identify which Talking Timer has alarmed when more than one timer is used. In the dark, in the clock mode, Talking Timer will announce the time while touching any button in the timer. There is also an option of turning the voice off.
Delivery information: Button cell batteries are included.
Specifications
- Description:Talking timer
- W×H×D:82.6×63½×25.4 mm (3¼×2½×1")
- Weight:0.06 kg (2.2 oz.)