Orders LinkContactUsLinkComponent
Aluminum Chucks, Electron Microscopy Sciences
Aluminum Chucks, Electron Microscopy Sciences
Abh-Jba/Aluminum Chucks
Catalog # 100492-892
Supplier #:  70174-20
Aluminum Chucks, Electron Microscopy Sciences
Catalog # 100492-892
Aluminum Chucks, Electron Microscopy Sciences
Aluminum Chucks, Electron Microscopy Sciences
Abh-Jba/Aluminum Chucks
Price
$77.03
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Machined from solid Aluminum
They come with set screws and a hex key.
Specifications
  • Description:
    Abh-Jba/Aluminum Chucks