Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Product Details & Documents
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.
- Each square grid fills one low-powered-field (approximately 100× magnification)
- Easy to locate an organism or cell every time
- Eighty one squares
The slides are available with coatings of Poly-L-Lysine or Silane, or they can be purchased plain (uncoated). The grid measures 2 mm square, 16 mm total. Numbers 1-8 and alpha A-H.
Specifications
- Description:Grid slides, uncoated