Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

Sodium Phosphate Buffer 2.0 M, 500 ml, Bottle, 7.4, Phosphate
Catalog # 100190-634
Marketsource
Supplier #:  26-516.5
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox
Catalog # 100190-634
Marketsource
CAS Number:  
Buffer Solution, Sodium Phosphate, pH 7.4, Moltox

Sodium Phosphate Buffer 2.0 M, 500 ml, Bottle, 7.4, Phosphate

CAS Number:  
Price
$132.35
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Sodium Phosphate Buffer 2.0 M
  • Size:
    500 ml
  • pH buffer type:
    Phosphate
  • pH:
    7.4

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.