$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MP-85 MANIPULATOR LEFT HANDED
100358-258
:
Sutter Instruments Company
MP-85 MANIPULATOR LEFT HANDED
100358-258
:
Sutter Instruments Company
:
MP-85/L