Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences

Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences

5.8 ml, 4.6×14.7 cm, Sterile, 25 µL, 3.3 ml
Catalog # 101413-324
Supplier #:  70966-12S
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences
Catalog # 101413-324
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences
Narrow Stem Pipettes - Sedi-Pet, Electron Microscopy Sciences

5.8 ml, 4.6×14.7 cm, Sterile, 25 µL, 3.3 ml

Price
$128.06
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These polyethylene disposable have a low-affinity surface, attached bulb avoids cross-contamination, and are one step biohazard disposable. These Micro-tip pipettes are ideal for loading gels, DNA sample removal, protein assays, lipids extractions, drop-wise additions to culture plates, using 2 ml auto sampler vials, remove or disperse small volumes, etc.

Specifications
  • Ø×L:
    4.6×14.7 cm
  • Sterility:
    Sterile
  • Volume:
    5.8 ml
  • Drop size:
    25 µL
  • Bulb draw:
    3.3 ml

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.