Orders LinkContactUsLinkComponent
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack
Catalog # 100502-460
Supplier #:  62015-01
Cryogenic Vial Rack, Electron Microscopy Sciences
Catalog # 100502-460
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack, Electron Microscopy Sciences
Cryogenic Vial Rack
Price
$55.24
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from durable polycarbonate. Its unique design with slotted cavities accepts vials with slotted skirts. When placed in the holes the vials become locked in position, enabling the user to attach or remove the cap with one hand. It accommodates up to 50 vials(1/2" or 13.5 mm diameter) in a 4x10 array with alphanumeric indexing. Autoclavable at 121°C. Measures: 190 x 108 x 22 mm (7 1/2 x 4 1/2 x 7/8").
Specifications
  • Description:
    Cryogenic Vial Rack