Orders LinkContactUsLinkComponent
Pen Vac™ Tool, Electron Microscopy Sciences
Pen Vac™ Tool, Electron Microscopy Sciences
Pen-Vac System, incl. One Pen with 6 Probes and Cups. (6 Probes: 3 angled,1/8", 1/4", 3/8" and 3 straight 1/8", 1/4", 3/8")
Catalog # 100501-340
Supplier #:  71914
Pen Vac™ Tool, Electron Microscopy Sciences
Catalog # 100501-340
Pen Vac™ Tool, Electron Microscopy Sciences
Pen Vac™ Tool, Electron Microscopy Sciences
Pen-Vac System, incl. One Pen with 6 Probes and Cups. (6 Probes: 3 angled,1/8", 1/4", 3/8" and 3 straight 1/8", 1/4", 3/8")
Price
$177.33
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pen-Vac™ is a new improved way to handle small, flat surface objects
  • Brushed aluminium body
  • Lifts upto 50 grams
  • Holds up to one minute
Pen-Vac™ comes with a variety of vacuum probes, complete with a vacuum cup attached. It is available with plastic or aluminum hubs and available in both straight and angled form. The stainless steel needle portion of the probes are one-half inch long. Vacuum Cups come in a wide range of materials. We offer the Static Dissipative and the Conductive Cups that provide ESD protection for electrostatic discharge of sensitive components. Cups comes in three sizes: 1/8" (3.17 mm); 1/4" (6.35 mm) and 3/8" (9.52 mm).
Beside the electronics assembly industry, jewelers, model builders etc. Pen-Vac is ideal for EM work as well. It can be used to handle grids, pick up stubs, align membranes, work with glass slides, cover slips and much more.
Specifications
  • Description:
    Pen-Vac System, incl. One Pen with 6 Probes and Cups. (6 Probes: 3 angled,1/8", 1/4", 3/8" and 3 straight 1/8", 1/4", 3/8")
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Pen-Vac Probe & Cup 1/8"
    Pen-Vac Probe & Cup 1/8"
    100501-962
    Log in for availability
    Pack of 2 - $13.12
Product Information
  • Catalog #
    Availability
    Price
  • Pen-Vac Probe & Cup¼"
    Pen-Vac Probe & Cup¼"
    100501-964
    Log in for availability
    Pack of 2 - $14.58
Product Information
  • Catalog #
    Availability
    Price
  • Pen-Vac Probe & Cup 3/8"
    Pen-Vac Probe & Cup 3/8"
    100501-966
    Log in for availability
    Pack of 2 - $14.58