Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences

Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences

Bowl size 15×15 mm, 15 cm
Catalog # 100493-950
Supplier #:  72984-15
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Catalog # 100493-950
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences

Bowl size 15×15 mm, 15 cm

Price
$68.72
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.

Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.

Specifications
  • Description:
    Bowl size 15×15 mm
  • Length:
    15 cm

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.