Orders LinkContactUsLinkComponent
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
  100491-748
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
  100491-748
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Grid Storage Box, 100 Grids, 8 L×5 W×7 T mm
$28.91
Each
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
  • Made from a special plastic that minimizes static
  • Complete with grid recording card
  • Capacity:
    100 Grids
  • Description:
    Grid Storage Box
  • Dimensions:
    8 L×5 W×7 T mm