Orders LinkContactUsLinkComponent
2-Ethoxyethanol ≥99%
250 ml
  101443-614
Marketsource
 :  1008401
2-Ethoxyethanol ≥99%
  101443-614
Marketsource
 :  110-80-5
2-Ethoxyethanol ≥99%
2-Ethoxyethanol ≥99%
250 ml
 :  110-80-5
$104.62
Each (250ml)
Use our stock solutions directly or as a component in solutions you create in your own lab. We prepare our stock solutions using high quality raw materials, ASTM Type 1 water, and sterile packaging. Our stock solutions are intended for Research and Development Use Only.
 :  Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve
  •  :110-80-5
  •  :C₂H₅OCH₂CH₂OH
  •  :MFCD00002869
  •  :90.12 g/mol
  •  :1171
  •  :3,III
  • Size:
    250 ml