Agar
Plastic bottle, 1 lbs.
Catalog # 101174-168
Marketsource
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Synonyms: Agar Agar
- CAS Number:9002-18-0
- MDL Number:MFCD00081288
- Merck Index:14,184
- Storage Temperature:Ambient
- EINECS:232-658-1
Specifications
Specification Test Results
- Total Ash:≤6.5%
- Loss on Drying:≤20.0%
- Identity:Passes Test
Specifications
- Pack type:Plastic bottle
- Size:1 lbs.
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



