Fixatives, Prepared, Electron Microscopy Sciences
Product Details & Documents
Premixed, prebuffered fixatives, that are pH adjusted for immediate laboratory use. They are freshly prepared upon receipt of order to ensure maximum shelf life. The fixatives are filled in 20 mL glass vials (10 mL/vial). This allows for the immediate fixation of your specimens without costly and time consuming transfer associated with ampoule contained fixatives.
Certifications: Certificates of analysis are available.
Documents
EMS-Core Sampling Tools
Product Literature
Specifications
Specifications
- Description:2.5% Glutaraldehyde in 0.1 M Millonigs Sodium Phosphate Buffer (pH 7.2)